missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Phenylpropyl)piperidine 98.0+%, TCI America™

Catalog No. P07605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07605G 5 g
P076025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07605G Supplier TCI America Supplier No. P07605G

Chemical Identifiers

CAS 18495-82-4
Molecular Formula C14H21N
Molecular Weight (g/mol) 203.33
MDL Number MFCD00023152
InChI Key HASRFXGIJALRRB-UHFFFAOYSA-N
PubChem CID 87678
IUPAC Name 4-(3-phenylpropyl)piperidine
SMILES C(CC1CCNCC1)CC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 203.33
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(3-Phenylpropyl)piperidine