missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Methylphenoxy)piperidine, 97%, Thermo Scientific Chemicals

Catalog No. AAH5095003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5095003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5095003 Supplier Thermo Scientific Chemicals Supplier No. H5095003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

4-(3-Methylphenoxy)piperidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(3-Methylphenoxy)piperidine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 63843-46-9
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD06248345
InChI Key RTSXWJCYCFVWBU-UHFFFAOYSA-N
Synonym 4-3-methylphenoxy piperidine, 4-m-tolyloxy piperidine, acmc-1b64o
PubChem CID 17196730
IUPAC Name 4-(3-methylphenoxy)piperidine
SMILES CC1=CC(=CC=C1)OC2CCNCC2

Specifications

Melting Point 190°C to 191°C
Quantity 1 g
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 4-(3-Methylphenoxy)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only