missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Hydroxypropyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™

Catalog No. H11415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11415G 5 g
H114125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11415G Supplier TCI America Supplier No. H11415G
Only null left

Chemical Identifiers

CAS 205194-33-8
Molecular Formula C7H15NO3S
Molecular Weight (g/mol) 193.261
InChI Key VAJSFWFPXNWBLF-UHFFFAOYSA-N
Synonym 3-(1,1-Dioxothiomorpholino)-1-propanol
PubChem CID 11435473
IUPAC Name 3-(1,1-dioxo-1,4-thiazinan-4-yl)propan-1-ol
SMILES C1CS(=O)(=O)CCN1CCCO

Specifications

Melting Point 72°C
Quantity 5 g
Formula Weight 193.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Hydroxypropyl)thiomorpholine 1,1-Dioxide