missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Hydroxyphenyl)piperidine 98.0+%, TCI America™

Catalog No. H09655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09655G 5 g
H096525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09655G Supplier TCI America Supplier No. H09655G

Chemical Identifiers

CAS 110878-71-2
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD02093474
InChI Key FINLIMGQGJZNRN-UHFFFAOYSA-N
Synonym 3-(4-Piperidyl)phenol
PubChem CID 10583453
IUPAC Name 3-piperidin-4-ylphenol
SMILES C1CNCCC1C2=CC(=CC=C2)O

Specifications

Melting Point 225°C
Color White-Yellow
Quantity 5 g
Formula Weight 177.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Hydroxyphenyl)piperidine