missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Fluorobenzyloxy)benzaldehyde 98.0+%, TCI America™

Catalog No. F09431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09431G 1 g
F09435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09431G Supplier TCI America Supplier No. F09431G

Chemical Identifiers

CAS 66742-57-2
Molecular Formula C14H11FO2
Molecular Weight (g/mol) 230.238
MDL Number MFCD02605317
InChI Key DNKSIIHRKWTIRH-UHFFFAOYSA-N
PubChem CID 880119
IUPAC Name 4-[(3-fluorophenyl)methoxy]benzaldehyde
SMILES C1=CC(=CC(=C1)F)COC2=CC=C(C=C2)C=O

Specifications

Melting Point 48°C
Color White
Boiling Point 265°C
Quantity 1 g
Formula Weight 230.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Fluorobenzyloxy)benzaldehyde