missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Dimethylamino)propoxy]benzonitrile, 97%, Thermo Scientific™

Catalog No. CC59516DE
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC59516DE 5g Amber Glass Bottle
CC59516DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC59516DE Supplier Thermo Scientific Chemicals Supplier No. CC59516DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 146440-15-5
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD09064993
InChI Key XYCJVOUBTBJYTL-UHFFFAOYSA-N
Synonym 4-3-dimethylamino propoxy benzonitrile, benzonitrile,4-3-dimethylamino propoxy, acmc-20n4ty, benzonitrile, 4-3-dimethylamino propoxy, 4-3-dimethylamino propoxy benzenecarbonitrile
PubChem CID 11858362
IUPAC Name 4-[3-(dimethylamino)propoxy]benzonitrile
SMILES CN(C)CCCOC1=CC=C(C=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 4-[3-(Dimethylamino)propoxy]benzonitrile
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.