missing translation for 'onlineSavingsMsg'
Learn More

4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline, 97%

Catalog No. p-7035993
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54229MD 250 mg
AAH5422903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54229MD Supplier Thermo Scientific Chemicals Supplier No. H54229MD
Only null left

CAS: 231278-20-9 | C21H14ClFIN3O | 505.72 g/mol

4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 231278-20-9
Molecular Formula C21H14ClFIN3O
Molecular Weight (g/mol) 505.72
MDL Number MFCD09998827
InChI Key UHFPFDMMKYQMLC-UHFFFAOYSA-N
Synonym n-3-chloro-4-3-fluorobenzyl oxy phenyl-6-iodoquinazolin-4-amine, n-3-chloro-4-3-fluorobenzyloxy phenyl-6-iodoquinazolin-4-amine, n-3-chloro-4-3-fluorobenzyloxy-phenyl-6-iodoquinazolin-4-amine, n-3-chloro-4-3-fluorophenyl methoxy phenyl-6-iodoquinazolin-4-amine, 6-iodo-4-3-chloro-4-3-fluorobenzyloxy anilino quinazoline, 4-3-chloro-4-3-fluorobenzyloxy phenylamino-6-iodoquinazoline, n-3-chloro-4-3-fluorobenzyl oxy phenyl-6-iodo-4-quinazolinamine, 4-quinazolinamine, n-3-chloro-4-3-fluorophenyl methoxy phenyl-6-iodo
PubChem CID 10174519
IUPAC Name N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-iodoquinazolin-4-amine
SMILES FC1=CC=CC(COC2=C(Cl)C=C(NC3=C4C=C(I)C=CC4=NC=N3)C=C2)=C1

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 505.72
Percent Purity 97%
Chemical Name or Material 4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only