missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Bromophenyl)pyridine 98.0+%, TCI America™

Catalog No. B46161G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46161G 1 g
B46165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46161G Supplier TCI America Supplier No. B46161G
Only null left

Chemical Identifiers

CAS 4373-72-2
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.10
MDL Number MFCD16250222
InChI Key LCTYNEHIFPECNL-UHFFFAOYSA-N
PubChem CID 13150402
IUPAC Name 4-(3-bromophenyl)pyridine
SMILES BrC1=CC=CC(=C1)C1=CC=NC=C1

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 1 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Bromophenyl)pyridine