missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3,4-Difluorophenyl)-1,3-thiazol-2-amine, 97%, Thermo Scientificâ„¢
Chemical Identifiers
CAS | 175135-32-7 |
---|---|
Molecular Formula | C9H6F2N2S |
Molecular Weight (g/mol) | 212.218 |
MDL Number | MFCD00052876 |
InChI Key | NDCSJUJQMRFHEX-UHFFFAOYSA-N |
Synonym | 4-3,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-3,4-difluorophenyl thiazole, 4-3,4-difluorophenyl thiazol-2-amine, 4-3,4-difluoro-phenyl-thiazol-2-ylamine, 2-amino-4-3,4-difluorophenyl-1,3-thiazole, 2-thiazolamine,4-3,4-difluorophenyl, 4-3,4-difluoro-phenyl-1,3-thiazol-2-amine, 4-3,4-difluorophenyl-1,3-thiazole-2-ylamine, pubchem14456, timtec-bb sbb005523 |
PubChem CID | 706326 |
IUPAC Name | 4-(3,4-difluorophenyl)-1,3-thiazol-2-amine |
SMILES | C1=CC(=C(C=C1C2=CSC(=N2)N)F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BTB07014DA
|
Thermo Scientific Chemicals
BTB07014DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB07014EA
|
Thermo Scientific Chemicals
BTB07014EA |
10g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
175135-32-7 | |
212.218 | |
NDCSJUJQMRFHEX-UHFFFAOYSA-N | |
706326 | |
C1=CC(=C(C=C1C2=CSC(=N2)N)F)F |
C9H6F2N2S | |
MFCD00052876 | |
4-3,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-3,4-difluorophenyl thiazole, 4-3,4-difluorophenyl thiazol-2-amine, 4-3,4-difluoro-phenyl-thiazol-2-ylamine, 2-amino-4-3,4-difluorophenyl-1,3-thiazole, 2-thiazolamine,4-3,4-difluorophenyl, 4-3,4-difluoro-phenyl-1,3-thiazol-2-amine, 4-3,4-difluorophenyl-1,3-thiazole-2-ylamine, pubchem14456, timtec-bb sbb005523 | |
4-(3,4-difluorophenyl)-1,3-thiazol-2-amine |
Specifications
175135-32-7 | |
100.0 | |
Amber Glass Bottle | |
MFCD00052876 | |
4-3,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-3,4-difluorophenyl thiazole, 4-3,4-difluorophenyl thiazol-2-amine, 4-3,4-difluoro-phenyl-thiazol-2-ylamine, 2-amino-4-3,4-difluorophenyl-1,3-thiazole, 2-thiazolamine,4-3,4-difluorophenyl, 4-3,4-difluoro-phenyl-1,3-thiazol-2-amine, 4-3,4-difluorophenyl-1,3-thiazole-2-ylamine, pubchem14456, timtec-bb sbb005523 | |
C1=CC(=C(C=C1C2=CSC(=N2)N)F)F | |
212.218 | |
212.22 | |
4-(3, 4-Difluorophenyl)-1, 3-thiazol-2-amine |
96.5 | |
97% | |
C9H6F2N2S | |
1g | |
NDCSJUJQMRFHEX-UHFFFAOYSA-N | |
4-(3,4-difluorophenyl)-1,3-thiazol-2-amine | |
706326 | |
97% |