missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine 98.0+%, TCI America™
Supplier: TCI America T33991G
Specifications
| 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine | |
| 99°C | |
| C17H20BNO2 | |
| 1 g | |
| ZTGKWVBDXOXURT-UHFFFAOYSA-N | |
| 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine | |
| 57416227 | |
| ≥98.0% (GC,T) |
| 1009033-83-3 | |
| White-Yellow | |
| MFCD20923942 | |
| 4,4,5,5-Tetramethyl-2-[3-(4-pyridyl)phenyl]-1,3,2-dioxaborolane, 3-(4-Pyridyl)phenylboronic Acid Pinacol Ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C3=CC=NC=C3 | |
| 281.162 | |
| 281.16 | |
| Crystalline Powder |
Chemical Identifiers
| 1009033-83-3 | |
| 281.162 | |
| ZTGKWVBDXOXURT-UHFFFAOYSA-N | |
| 57416227 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C3=CC=NC=C3 |
| C17H20BNO2 | |
| MFCD20923942 | |
| 4,4,5,5-Tetramethyl-2-[3-(4-pyridyl)phenyl]-1,3,2-dioxaborolane, 3-(4-Pyridyl)phenylboronic Acid Pinacol Ester | |
| 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine |
Safety and Handling
TSCA : No