missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine 98.0+%, TCI America™

Numéro de catalogue. T3399200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T3399200MG 200 mg
T33991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T3399200MG Fournisseur TCI America Code fournisseur T3399200MG
Il en reste null

Identifiants chimiques

CAS 1009033-83-3
Molecular Formula C17H20BNO2
Molecular Weight (g/mol) 281.162
MDL Number MFCD20923942
InChI Key ZTGKWVBDXOXURT-UHFFFAOYSA-N
Synonym 4,4,5,5-Tetramethyl-2-[3-(4-pyridyl)phenyl]-1,3,2-dioxaborolane, 3-(4-Pyridyl)phenylboronic Acid Pinacol Ester
PubChem CID 57416227
IUPAC Name 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C3=CC=NC=C3

Spécifications

Melting Point 99°C
Color White-Yellow
Quantity 200 mg
Formula Weight 281.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine