missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Trimethylsilyl)ethoxycarbonyloxy]nitrobenzene 98.0+%, TCI America™

Catalog No. T28725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28725G Supplier TCI America Supplier No. T28725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80149-80-0
Molecular Formula C12H17NO5Si
Molecular Weight (g/mol) 283.36
MDL Number MFCD00042930
InChI Key ZAQWGGKIMQIVGM-UHFFFAOYSA-N
Synonym 4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204
PubChem CID 3086116
IUPAC Name 4-nitrophenyl 2-(trimethylsilyl)ethyl carbonate
SMILES C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 39°C
Color Yellow
Quantity 5 g
Formula Weight 283.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[2-(Trimethylsilyl)ethoxycarbonyloxy]nitrobenzene
TSCA No
Recommended Storage Refrigerator