missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%

Catalog No. p-3779535
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC176320050 5 g Glass bottle
AC176320250 25 g Glass bottle
AC176321000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC176320050 Supplier Thermo Scientific Chemicals Supplier No. 176320050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16593-81-0 | C11H9N3O2 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Specifications

Chemical Name or Material 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
CAS 16593-81-0, 1141-59-9
CAS Min % 99.0
CAS Max % 100.0
Color Brown to Brown-Orange
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C11H9N3O2
Quantity 5 g
MDL Number MFCD00150712,MFCD00150712,MFCD00150712,MFCD00006256
Beilstein 22, I, 694
Merck Index 15, 7125
Synonym 4-2-pyridylazo resorcinol monosodium saltmonohydrate
Solubility Information (0.1% in water) Clear
InChI Key VLCAILLZPUINNF-UKTHLTGXSA-N
SMILES OC1=CC(=O)C=C\C1=N/NC1=CC=CC=N1
IUPAC Name 3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one;sodium;hydrate
Molecular Weight (g/mol) 215.21
PubChem CID 132988475
Formula Weight 255.21
Percent Purity 99+%
Physical Form Crystalline Powder
Show More Show Less

RUO – Research Use Only