Learn More
4-(2-Pyridylazo)resorcinol, ACS
CAS: 1141-59-9 | C11H9N3O2 | 215.212 g/mol
$60.55 - $165.22
Chemical Identifiers
| CAS | 1141-59-9 |
|---|---|
| Molecular Formula | C11H9N3O2 |
| Molecular Weight (g/mol) | 215.212 |
| MDL Number | MFCD00006256 |
| InChI Key | VLCAILLZPUINNF-LCYFTJDESA-N |
| Synonym | 4-2-pyridylazo resorcinol, 4-2-pyridlyazo resorcinol, 4-2-pyridylazo-2-resorcinol, resorcinol, 4-2-pyridylazo, 1-2-pyridylazo resorcinol, unii-n68t40h95t, 1,3-benzenediol, 4-2-pyridinylazo, 4-pyridin-2-yldiazenyl benzene-1,3-diol, 4-2-pyridylazo-1,3-benzenediol, 1,3-benzenediol, 4-2-2-pyridinyl diazenyl |
| PubChem CID | 5474737 |
| IUPAC Name | (4Z)-3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one |
| SMILES | C1=CC=NC(=C1)NN=C2C=CC(=O)C=C2O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1141-59-9 | |
| 215.212 | |
| VLCAILLZPUINNF-LCYFTJDESA-N | |
| 5474737 | |
| C1=CC=NC(=C1)NN=C2C=CC(=O)C=C2O |
| C11H9N3O2 | |
| MFCD00006256 | |
| 4-2-pyridylazo resorcinol, 4-2-pyridlyazo resorcinol, 4-2-pyridylazo-2-resorcinol, resorcinol, 4-2-pyridylazo, 1-2-pyridylazo resorcinol, unii-n68t40h95t, 1,3-benzenediol, 4-2-pyridinylazo, 4-pyridin-2-yldiazenyl benzene-1,3-diol, 4-2-pyridylazo-1,3-benzenediol, 1,3-benzenediol, 4-2-2-pyridinyl diazenyl | |
| (4Z)-3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one |
Specifications
| 1141-59-9 | |
| C11H9N3O2 | |
| 1 g | |
| 4-2-pyridylazo resorcinol, 4-2-pyridlyazo resorcinol, 4-2-pyridylazo-2-resorcinol, resorcinol, 4-2-pyridylazo, 1-2-pyridylazo resorcinol, unii-n68t40h95t, 1,3-benzenediol, 4-2-pyridinylazo, 4-pyridin-2-yldiazenyl benzene-1,3-diol, 4-2-pyridylazo-1,3-benzenediol, 1,3-benzenediol, 4-2-2-pyridinyl diazenyl | |
| C1=CC=NC(=C1)NN=C2C=CC(=O)C=C2O | |
| 215.212 | |
| 215.21 | |
| Powder |
| 187°C (decomposition) | |
| MFCD00006256 | |
| 184665 | |
| VLCAILLZPUINNF-LCYFTJDESA-N | |
| (4Z)-3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one | |
| 5474737 | |
| ACS | |
| 4- (2-Pyridylazo)resorcinol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-528-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only