missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyridylazo)resorcinol, 97+%

Catalog No. AC418590050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418590050 5 g Glass bottle
AC418590250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418590050 Supplier Thermo Scientific Chemicals Supplier No. 418590050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1141-59-9 | C11H9N3O2 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 1141-59-9
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.21
MDL Number MFCD00006256
InChI Key VLCAILLZPUINNF-LCYFTJDESA-N
Synonym 4-2-pyridylazo resorcinol, 4-2-pyridlyazo resorcinol, 4-2-pyridylazo-2-resorcinol, resorcinol, 4-2-pyridylazo, 1-2-pyridylazo resorcinol, unii-n68t40h95t, 1,3-benzenediol, 4-2-pyridinylazo, 4-pyridin-2-yldiazenyl benzene-1,3-diol, 4-2-pyridylazo-1,3-benzenediol, 1,3-benzenediol, 4-2-2-pyridinyl diazenyl
PubChem CID 5474737
IUPAC Name (4Z)-3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES C1=CC=NC(=C1)NN=C2C=CC(=O)C=C2O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 188.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 7125
Formula Weight 215.21
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 4-(2-Pyridylazo)resorcinol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-528-6

RUO – Research Use Only