missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Propynyloxy)aniline 98.0+%, TCI America™

Catalog No. p-7135465
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22241G 1 g
P22245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22241G Supplier TCI America Supplier No. P22241G

Chemical Identifiers

CAS 26557-78-8
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD12823190
InChI Key FPLSLCJCSKFAMA-UHFFFAOYSA-N
Synonym 4-(Propargyloxy)aniline, 4-Aminophenyl Propargyl Ether
PubChem CID 12668862
IUPAC Name 4-(prop-2-yn-1-yloxy)aniline
SMILES NC1=CC=C(OCC#C)C=C1

Specifications

Melting Point 54°C
Color White
Boiling Point 95°C
Quantity 1 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Propynyloxy)aniline
TSCA No
Recommended Storage Refrigerator