missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride, 97%

CAS: 112539-09-0 | C10H10ClNO3S | 259.704 g/mol

$106.63 - $940.90

Chemical Identifiers

CAS 112539-09-0
Molecular Formula C10H10ClNO3S
Molecular Weight (g/mol) 259.704
MDL Number MFCD00219329
InChI Key OUAVTZYGHGCCCB-UHFFFAOYSA-N
PubChem CID 2794593
IUPAC Name 4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride
SMILES C1CC(=O)N(C1)C2=CC=C(C=C2)S(=O)(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH34441MD
View Documents
Thermo Scientific Chemicals
H34441MD
250 mg N/A
Only null left
 
AAH3444103
View Documents
Thermo Scientific Chemicals
H3444103
1 g N/A
Only null left
 
AAH3444106
View Documents
Thermo Scientific Chemicals
H3444106
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112539-09-0
259.704
OUAVTZYGHGCCCB-UHFFFAOYSA-N
4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride
C10H10ClNO3S
MFCD00219329
2794593
C1CC(=O)N(C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

112539-09-0
C10H10ClNO3S
250 mg
Air and moisture sensitive
OUAVTZYGHGCCCB-UHFFFAOYSA-N
4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride
2794593
97%
104°C to 105°C
MFCD00219329
UN3261
4-2-oxopyrrolidin-1-yl benzenesulfonyl chloride, 4-2-oxo-1-pyrrolidinyl benzenesulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonylchloride, 4-2-oxopyrrolidin-1-yl benzene-1-sulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonyl chloride, benzenesulfonylchloride, 4-2-oxo-1-pyrrolidinyl, 1-4-chlorosulfonyl phenyl pyrrolidin-2-one, pubchem5680, acmc-1bri4, benzenesulfonylchloride,4-2-oxo-1-pyrrolidinyl
C1CC(=O)N(C1)C2=CC=C(C=C2)S(=O)(=O)Cl
259.704
259.71
4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only