missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Octylamino)diphenylamine 98.0+%, TCI America™
$29.83 - $277.75
Chemical Identifiers
| CAS | 15233-47-3 |
|---|---|
| Molecular Formula | C20H28N2 |
| Molecular Weight (g/mol) | 296.458 |
| MDL Number | MFCD00059441 |
| InChI Key | JQTYAZKTBXWQOM-UHFFFAOYSA-N |
| PubChem CID | 85821 |
| IUPAC Name | 4-N-octan-2-yl-1-N-phenylbenzene-1,4-diamine |
| SMILES | CCCCCCC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2 |
Chemical Identifiers
| 15233-47-3 | |
| 296.458 | |
| JQTYAZKTBXWQOM-UHFFFAOYSA-N | |
| 4-N-octan-2-yl-1-N-phenylbenzene-1,4-diamine |
| C20H28N2 | |
| MFCD00059441 | |
| 85821 | |
| CCCCCCC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2 |
Specifications
| 15233-47-3 | |
| MFCD00059441 | |
| N-(1-Methylheptyl)-N′C-phenyl-1,4-phenylenediamine, N-Phenyl-N′C-(2-octyl)-1,4-phenylenediamine | |
| CCCCCCC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2 | |
| 296.458 | |
| 296.46 | |
| Liquid |
| C20H28N2 | |
| 25 g | |
| JQTYAZKTBXWQOM-UHFFFAOYSA-N | |
| 4-N-octan-2-yl-1-N-phenylbenzene-1,4-diamine | |
| 85821 | |
| ≥98.0% (GC,T) | |
| 4-(2-Octylamino)diphenylamine |
Safety and Handling
EINECSNumber : (3)-0136&(3)-0368
TSCA : Yes