missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Octylamino)diphenylamine 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 500 g |
| Synonym | N-(1-Methylheptyl)-N′-phenyl-1,4-phenylenediamine, N-Phenyl-N′-(2-octyl)-1,4-phenylenediamine |
| Formula Weight | 296.46 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-(2-Octylamino)diphenylamine |
Safety and Handling
| EINECSNumber | (3)-0136&(3)-0368 |
| TSCA | Yes |