missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Octylamino)diphenylamine 98.0+%, TCI America™

Catalog No. O016625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O016625G 25 g
O0166500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O016625G Supplier TCI America Supplier No. O016625G
Only null left

Chemical Identifiers

CAS 15233-47-3
Molecular Formula C20H28N2
Molecular Weight (g/mol) 296.458
MDL Number MFCD00059441
InChI Key JQTYAZKTBXWQOM-UHFFFAOYSA-N
PubChem CID 85821
IUPAC Name 4-N-octan-2-yl-1-N-phenylbenzene-1,4-diamine
SMILES CCCCCCC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

Quantity 25 g
Synonym N-(1-Methylheptyl)-N′C-phenyl-1,4-phenylenediamine, N-Phenyl-N′C-(2-octyl)-1,4-phenylenediamine
Formula Weight 296.46
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(2-Octylamino)diphenylamine
EINECSNumber (3)-0136&(3)-0368
TSCA Yes