missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(N,N-Dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015898
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015898 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015898 Supplier Merck Emd Millipore Supplier No. 79291100MG

Chemical Identifiers

MDL Number MFCD21363128
Synonym 4-[2-(N,N-Dimethylamino)ethylsulfamoyl]-7-(2-aminoethylamino)benzofurazan; 7-[(2-Aminoethyl)amino]-N-[2-(dimethylamino)ethyl]-2,1,3-benzoxadiazole-4-sulfonamide; DAABD-AE

Specifications

CAS 913253-56-2
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C12H20N6O3 S
Quantity 100 mg
Formula Weight 328.39
Percent Purity ≥95.0% (HPLC)
Grade For HPLC derivatization
ShelfLife Limited shelf life, expiry date on the label