missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Methyl-4-thiazolyl)phenol 98.0+%, TCI America™
$104.38 - $380.27
Chemical Identifiers
| CAS | 30686-73-8 |
|---|---|
| Molecular Formula | C10H9NOS |
| Molecular Weight (g/mol) | 191.248 |
| MDL Number | MFCD02642878 |
| InChI Key | HOAYTTLLVORNLA-UHFFFAOYSA-N |
| Synonym | 4-2-methyl-4-thiazolyl phenol, 4-2-methyl-1,3-thiazol-4-yl phenol, 4-2-methylthiazol-4-yl phenol, 4-4-hydroxyphenyl-2-methylthiazole, 4-2-methyl-4-thiazolyl phenol for biochemical research, 4-2-methyl-thiazol-4-yl-phenol, phenol, p-2-methyl-4-thiazolyl, 4-2-methyl-4-thiazolyl phenol*, 4-2-methyl-1,3-thiazol-4-yl phenol #, phenol, 4-2-methyl-4-thiazolyl-, hydrochloride |
| PubChem CID | 591743 |
| IUPAC Name | 4-(2-methyl-1,3-thiazol-4-yl)phenol |
| SMILES | CC1=NC(=CS1)C2=CC=C(C=C2)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M2325100MG
|
TCI America
M2325100MG |
100 mg |
Each for $104.38
|
|
|||||
|
M23251G
|
TCI America
M23251G |
1 g |
Each for $380.27
|
|
|||||
Chemical Identifiers
| 30686-73-8 | |
| 191.248 | |
| HOAYTTLLVORNLA-UHFFFAOYSA-N | |
| 591743 | |
| CC1=NC(=CS1)C2=CC=C(C=C2)O |
| C10H9NOS | |
| MFCD02642878 | |
| 4-2-methyl-4-thiazolyl phenol, 4-2-methyl-1,3-thiazol-4-yl phenol, 4-2-methylthiazol-4-yl phenol, 4-4-hydroxyphenyl-2-methylthiazole, 4-2-methyl-4-thiazolyl phenol for biochemical research, 4-2-methyl-thiazol-4-yl-phenol, phenol, p-2-methyl-4-thiazolyl, 4-2-methyl-4-thiazolyl phenol*, 4-2-methyl-1,3-thiazol-4-yl phenol #, phenol, 4-2-methyl-4-thiazolyl-, hydrochloride | |
| 4-(2-methyl-1,3-thiazol-4-yl)phenol |
Specifications
| 30686-73-8 | |
| White-Yellow | |
| MFCD02642878 | |
| 4-2-methyl-4-thiazolyl phenol, 4-2-methyl-1,3-thiazol-4-yl phenol, 4-2-methylthiazol-4-yl phenol, 4-4-hydroxyphenyl-2-methylthiazole, 4-2-methyl-4-thiazolyl phenol for biochemical research, 4-2-methyl-thiazol-4-yl-phenol, phenol, p-2-methyl-4-thiazolyl, 4-2-methyl-4-thiazolyl phenol*, 4-2-methyl-1,3-thiazol-4-yl phenol #, phenol, 4-2-methyl-4-thiazolyl-, hydrochloride | |
| CC1=NC(=CS1)C2=CC=C(C=C2)O | |
| 191.248 | |
| 191.25 | |
| Crystalline Powder |
| 213°C | |
| C10H9NOS | |
| 100 mg | |
| HOAYTTLLVORNLA-UHFFFAOYSA-N | |
| 4-(2-methyl-1,3-thiazol-4-yl)phenol | |
| 591743 | |
| ≥98.0% (HPLC,N) | |
| 4-(2-Methyl-4-thiazolyl)phenol [for Biochemical Research] |
Safety and Handling
TSCA : No