missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Méthyl-1H-imidazol-1-yl)benzylamine, 90%, Thermo Scientific™

Numéro de catalogue. CC61413DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC61413DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC61413DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC61413DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 883291-45-0
Formule moléculaire C11H13N3
Poids moléculaire (g/mol) 187.246
Numéro MDL MFCD06617944
Clé InChI JOMOQABDRBJMDQ-UHFFFAOYSA-N
Synonyme 4-2-methyl-1h-imidazol-1-yl benzylamine, 1-4-2-methyl-1h-imidazol-1-yl phenyl methanamine, 4-2-methylimidazol-1-yl phenyl methanamine, benzenemethanamine,4-2-methyl-1h-imidazol-1-yl, 4-2-methyl-1h-imidazol-1-yl phenyl methanamine, 4-2-methylimidazolyl phenyl methylamine, 1-4-2-methylimidazol-1-yl phenyl methanamine
PubChem CID 4913134
Nom de l’IUPAC [4-(2-méthylimidazol-1-yl)phényl]méthanamine
SOURIRES CC1=NC=CN1C2=CC=C(C=C2)CN

Spécifications

Nom ou substance chimique 4-(2-Methyl-1H-imidazol-1-yl)benzylamine
CAS % minimum 89.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 90%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 187.24
Pourcentage de pureté 90%
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.