missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-1H-imidazol-1-yl)benzoic acid hydrochloride hydrate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC61451DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC61451DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

921938-78-5
256.686
4-2-methyl-1h-imidazol-1-yl benzoic acid hydrochloride hydrate, 4-2-methylimidazol-1-yl benzoic acid hydrate hydrochloride
4-(2-methylimidazol-1-yl)benzoic acid;hydrate;hydrochloride
C11H13ClN2O3
PSHJNJCAKINUEP-UHFFFAOYSA-N
44118781
CC1=NC=CN1C2=CC=C(C=C2)C(=O)O.O.Cl

Specifications

4-(2-Methyl-1H-imidazol-1-yl)benzoic acid hydrochloride hydrate
95%
C11H13ClN2O3
4-2-methyl-1h-imidazol-1-yl benzoic acid hydrochloride hydrate, 4-2-methylimidazol-1-yl benzoic acid hydrate hydrochloride
CC1=NC=CN1C2=CC=C(C=C2)C(=O)O.O.Cl
256.686
256.69
921938-78-5
Amber Glass Bottle
5g
PSHJNJCAKINUEP-UHFFFAOYSA-N
4-(2-methylimidazol-1-yl)benzoic acid;hydrate;hydrochloride
44118781
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents