missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester, 97%

Catalog No. AAH2794503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2794503 1 g
AAH2794506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2794503 Supplier Thermo Scientific Chemicals Supplier No. H2794503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1073353-60-2 | C16H24BNO4 | 305.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073353-60-2
Molecular Formula C16H24BNO4
Molecular Weight (g/mol) 305.18
MDL Number MFCD09266188
InChI Key XNDAIQVEKVZONM-UHFFFAOYSA-N
Synonym n-2-methoxyethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-2-methoxyethylcarbamoyl phenylboronic acid, pinacol ester, 4-2-methoxyethylamine-1-carbonyl phenyl boronic acid pinacol ester, amtb042, 4-2-methoxyethyl carbamoyl phenylboronic acid pinacol ester, 4-2-methoxyethylaminecarbonyl benzeneboronic acid pinacol ester, n-2-methoxyethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
PubChem CID 46739044
IUPAC Name N-(2-methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES COCCNC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 73°C to 76°C
Quantity 1 g
Formula Weight 305.18
Percent Purity 97%
Chemical Name or Material 4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only