missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Hydroxyethyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™
Supplier: TCI America H114025G
Specifications
| 4-(2-Hydroxyethyl)thiomorpholine 1,1-Dioxide | |
| 74°C | |
| 25 g | |
| GILLKIJFJOBJCD-UHFFFAOYSA-N | |
| 2-(1,1-dioxo-1,4-thiazinan-4-yl)ethanol | |
| 21874811 | |
| ≥98.0% (GC,T) |
| 26475-62-7 | |
| C6H13NO3S | |
| 2-(1,1-Dioxothiomorpholino)ethanol | |
| C1CS(=O)(=O)CCN1CCO | |
| 179.234 | |
| 179.23 | |
| Crystalline Powder |
Chemical Identifiers
| 26475-62-7 | |
| 179.234 | |
| 2-(1,1-Dioxothiomorpholino)ethanol | |
| 2-(1,1-dioxo-1,4-thiazinan-4-yl)ethanol |
| C6H13NO3S | |
| GILLKIJFJOBJCD-UHFFFAOYSA-N | |
| 21874811 | |
| C1CS(=O)(=O)CCN1CCO |
Safety and Handling
TSCA : No