missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™

Catalog No. H11405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11405G 5 g
H114025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11405G Supplier TCI America Supplier No. H11405G

Chemical Identifiers

CAS 26475-62-7
Molecular Formula C6H13NO3S
Molecular Weight (g/mol) 179.234
InChI Key GILLKIJFJOBJCD-UHFFFAOYSA-N
Synonym 2-(1,1-Dioxothiomorpholino)ethanol
PubChem CID 21874811
IUPAC Name 2-(1,1-dioxo-1,4-thiazinan-4-yl)ethanol
SMILES C1CS(=O)(=O)CCN1CCO

Specifications

Melting Point 74°C
Quantity 5 g
Formula Weight 179.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Hydroxyethyl)thiomorpholine 1,1-Dioxide