missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)pyridine, 98%

Catalog No. AAL1378006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1378006 5 g
AAL1378003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1378006 Supplier Thermo Scientific Chemicals Supplier No. L1378006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5344-27-4 | C7H9NO | 123.155 g/mol

4-(2-Hydroxyethyl)pyridine acts as a reagent for the protection of carboxyl groups as their 2-(4-pyridyl)ethyl esters in organic synthesis. Further, it is used as an active pharmaceutical ingredient intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Hydroxyethyl)pyridine acts as a reagent for the protection of carboxyl groups as their 2-(4-pyridyl)ethyl esters in organic synthesis. Further, it is used as an active pharmaceutical ingredient intermediate.

Solubility
Miscible with water.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents, bases and mineral acids.

Chemical Identifiers

CAS 5344-27-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00129038
InChI Key DWPYQDGDWBKJQL-UHFFFAOYSA-N
Synonym 4-pyridineethanol, 2-pyridin-4-yl ethanol, 4-2-hydroxyethyl pyridine, 4-ethanolpyridine, 2-4-pyridyl ethanol, 2-pyridin-4-yl ethan-1-ol, 2-pyridin-4-yl-ethanol, 2-gamma-pyridyl ethanol, unii-i89rdq5yd9
PubChem CID 72921
IUPAC Name 2-pyridin-4-ylethanol
SMILES C1=CN=CC=C1CCO

Specifications

Melting Point 8°C to 10°C
Density 1.093
Boiling Point 121°C to 122°C (2 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.538
Quantity 5 g
Beilstein 111361
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 4-(2-Hydroxyethyl)pyridine
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 226-286-9
RTECSNumber UT2971000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only