missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)piperazine-1-(2-hydroxypropane-3-sulfonic Acid) Hydrate 98.0+%, TCI America™

Catalog No. H077225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H077225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H077225G Supplier TCI America Supplier No. H077225G

[Good's buffer component for biological research]

Chemical Identifiers

CAS 68399-78-0
Molecular Formula C9H20N2O5S
Molecular Weight (g/mol) 268.328
MDL Number MFCD00038351
InChI Key GIZQLVPDAOBAFN-UHFFFAOYSA-N
Synonym heppso, 2-hydroxy-3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid, n-hydroxyethyl piperazine-n'-2-hydroxypropanesulfonic acid, beta-hydroxy-4-2-hydroxyethyl piperazine-1-propanesulphonic acid, beta-hydroxy-4-2-hydroxyethyl-1-piperazinepropanesulfonic acid, 3-4-2-hydroxyethyl-1-piperazinyl-2-hydroxypropanesulfonic acid, 4-2-hydroxyethyl piperazine-1-2-hydroxypropane-3-sulfonic acid, 4-2-hydroxyethyl piperazine-1-2-hydroxypropane sulfonic acid free acid, 2-hydroxy-3-4-2-hydroxyethyl piperazinyl propanesulfonic acid
PubChem CID 100205
ChEBI CHEBI:32951
IUPAC Name 2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCO)CC(CS(=O)(=O)O)O

Specifications

Color White
Quantity 25 g
Formula Weight 268.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Hydroxyethyl)piperazine-1-(2-hydroxypropane-3-sulfonic Acid) Hydrate [Good′s buffer compo