missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Hydroxyethyl)morpholine 99.0+%, TCI America™
$40.43 - $182.89
Chemical Identifiers
| CAS | 622-40-2 |
|---|---|
| Molecular Formula | C6H13NO2 |
| Molecular Weight (g/mol) | 131.175 |
| MDL Number | MFCD00006180 |
| InChI Key | KKFDCBRMNNSAAW-UHFFFAOYSA-N |
| Synonym | 2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, n-2-hydroxyethyl morpholine, 2-4-morpholinyl ethanol, morpholine ethanol, morpholineethanol, hydroxyethylmorpholine, ethanol, 2-morpholino, beta-morpholinoethanol |
| PubChem CID | 61163 |
| ChEBI | CHEBI:67144 |
| IUPAC Name | 2-morpholin-4-ylethanol |
| SMILES | C1COCCN1CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H024425ML
|
TCI America
H024425ML |
25 mL |
Each for $40.43
|
|
|||||
|
H0244100ML
|
TCI America
H0244100ML |
100 mL |
Each for $59.69
|
|
|||||
|
H0244500ML
|
TCI America
H0244500ML |
500 mL |
Each for $182.89
|
|
|||||
Chemical Identifiers
| 622-40-2 | |
| 131.175 | |
| KKFDCBRMNNSAAW-UHFFFAOYSA-N | |
| 61163 | |
| 2-morpholin-4-ylethanol |
| C6H13NO2 | |
| MFCD00006180 | |
| 2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, n-2-hydroxyethyl morpholine, 2-4-morpholinyl ethanol, morpholine ethanol, morpholineethanol, hydroxyethylmorpholine, ethanol, 2-morpholino, beta-morpholinoethanol | |
| CHEBI:67144 | |
| C1COCCN1CCO |
Specifications
| 622-40-2 | |
| Yellow | |
| 222°C | |
| MFCD00006180 | |
| 2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, n-2-hydroxyethyl morpholine, 2-4-morpholinyl ethanol, morpholine ethanol, morpholineethanol, hydroxyethylmorpholine, ethanol, 2-morpholino, beta-morpholinoethanol | |
| C1COCCN1CCO | |
| 131.175 | |
| CHEBI:67144 | |
| ≥99.0% (GC) | |
| 4-(2-Hydroxyethyl)morpholine |
| 2°C | |
| 10.7 (100g/L H2O 20°C) | |
| C6H13NO2 | |
| 25 mL | |
| KKFDCBRMNNSAAW-UHFFFAOYSA-N | |
| 2-morpholin-4-ylethanol | |
| 61163 | |
| 131.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-6876
RTECSNumber : QE3510000
TSCA : Yes
Recommended Storage : Refrigerator