missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Hydroxyethyl)morpholine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 2°C |
| Color | Yellow |
| pH | 10.7 (100g/L H2O 20°C) |
| Boiling Point | 222°C |
| Quantity | 500 mL |
| Formula Weight | 131.18 |
| Percent Purity | ≥99.0% (GC) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | 4-(2-Hydroxyethyl)morpholine |
Safety and Handling
| EINECSNumber | (5)-6876 |
| RTECSNumber | QE3510000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |