missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Furyl)but-3-en-2-one, 97%, Thermo Scientific™

Catalog No. p-2700556
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD11326EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CD11326EA Supplier Thermo Scientific Chemicals Supplier No. CD11326EA
Only null left

Chemical Identifiers

CAS 623-15-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00039566
InChI Key GBKGJMYPQZODMI-SNAWJCMRSA-N
Synonym furfurylideneacetone, 4-2-furyl-3-buten-2-one, 3-buten-2-one, 4-2-furanyl, fam polymer, oramin r, furfural acetone, oramin special gr, furfuralacetone polymer, polyfurfurylidineacetone, furfuralacetone
PubChem CID 735940
IUPAC Name (E)-4-(furan-2-yl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material 4-(2-Furyl)but-3-en-2-one
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.