missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Furyl)benzoic acid, 97%, Thermo Scientific™

Catalog No. CC34401CB
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC34401CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC34401CB Supplier Thermo Scientific Chemicals Supplier No. CC34401CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 35461-98-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H8O3
Quantity 250mg
MDL Number MFCD04039073
Synonym 4-2-furyl benzoic acid, 4-furan-2-yl benzoic acid, 4-furan-2-yl-benzoic acid, 2-4-carboxyphenyl furan, 4-fur-2-ylbenzoic acid, p-2-furyl benzoic acid, 4-2-furanyl benzoic acid
InChI Key FOJYVBSPOBUCMV-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=C(C=C1)C1=CC=CO1
IUPAC Name 4-(furan-2-yl)benzoic acid
Molecular Weight (g/mol) 188.18
PubChem CID 5138791
Formula Weight 188.18
Percent Purity 97%
Chemical Name or Material 4-(2-Furyl)benzoic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.