missing translation for 'onlineSavingsMsg'
Learn More

4-(2-furyl)benzaldehyde, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC34404DE

Quantity 5g

 View more versions of this product

Catalog No. CC34404DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

60456-77-1
172.183
4-2-furyl benzaldehyde, 4-furan-2-yl benzaldehyde, 4-furan-2-yl-benzaldehyde, benzaldehyde,4-2-furanyl, 2-4-formylphenyl furan, 4-2-furanyl benzaldehyde
4-(furan-2-yl)benzaldehyde
C11H8O2
WBUXKMOCVYRVES-UHFFFAOYSA-N
2772295
C1=COC(=C1)C2=CC=C(C=C2)C=O

Specifications

4-(2-furyl)benzaldehyde
Amber Glass Bottle
5g
WBUXKMOCVYRVES-UHFFFAOYSA-N
4-(furan-2-yl)benzaldehyde
2772295
97%
60456-77-1
C11H8O2
4-2-furyl benzaldehyde, 4-furan-2-yl benzaldehyde, 4-furan-2-yl-benzaldehyde, benzaldehyde,4-2-furanyl, 2-4-formylphenyl furan, 4-2-furanyl benzaldehyde
C1=COC(=C1)C2=CC=C(C=C2)C=O
172.183
172.18
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents