missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Furyl)-3-buten-2-one 98.0+%, TCI America™

Catalog No. p-7129561
Change view
Click to view available options
Quantity:
100 g
25 g
Catalog No. Quantity
F023525G 25 g
F0235100G 100 g
2 options

Catalog No. F023525G

Supplier: TCI America F023525G

Only null left

Chemical Identifiers

CAS 623-15-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00039566
InChI Key GBKGJMYPQZODMI-SNAWJCMRSA-N
Synonym furfurylideneacetone, 4-2-furyl-3-buten-2-one, 3-buten-2-one, 4-2-furanyl, fam polymer, oramin r, furfural acetone, oramin special gr, furfuralacetone polymer, polyfurfurylidineacetone, furfuralacetone
PubChem CID 735940
IUPAC Name (E)-4-(furan-2-yl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC=CO1

Specifications

Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Furyl)-3-buten-2-one
RTECSNumber EM9939000
TSCA Yes
Recommended Storage Refrigerator