missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Furyl)-3-buten-2-one, cis + trans, 98%

Catalog No. AAA1945606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1945606 5 g
AAA1945614 25 g
AAA1945622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1945606 Supplier Thermo Scientific Chemicals Supplier No. A1945606

CAS: 623-15-4 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 623-15-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00039566
InChI Key GBKGJMYPQZODMI-SNAWJCMRSA-N
Synonym furfurylideneacetone, 4-2-furyl-3-buten-2-one, 3-buten-2-one, 4-2-furanyl, fam polymer, oramin r, furfural acetone, oramin special gr, furfuralacetone polymer, polyfurfurylidineacetone, furfuralacetone
PubChem CID 735940
IUPAC Name (E)-4-(furan-2-yl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC=CO1

Specifications

Melting Point 33°C to 38°C
Boiling Point 112°C to 115°C (10 mmHg)
Flash Point 103°C (217°F)
Quantity 5 g
Beilstein 109696
Sensitivity Light sensitive
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 4-(2-Furyl)-3-buten-2-one, cis + trans
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-774-3
RTECSNumber EM9939000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only