missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Fluorobenzyl)piperidine hydrochloride, 97%

Catalog No. p-7055307
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6384903 1 g
AAH6384906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6384903 Supplier Thermo Scientific Chemicals Supplier No. H6384903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 193357-21-0 | C12H17ClFN | 229.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 193357-21-0
Molecular Formula C12H17ClFN
Molecular Weight (g/mol) 229.72
MDL Number MFCD08669673
InChI Key XZRQMBCQGGXDKQ-UHFFFAOYSA-N
Synonym 4-3-fluorobenzyl piperidine hydrochloride, 4-3-fluorophenyl methyl piperidine hydrochloride, 4-3-fluoro-benzyl-piperidine hydrochloride, 4-3-fluorobenzyl piperidine hcl
PubChem CID 17039493
SMILES Cl.FC1=CC=CC(CC2CCNCC2)=C1

Specifications

Color White
Quantity 1 g
IUPAC Name 4-[(3-fluorophenyl)methyl]piperidine hydrochloride
Formula Weight 229.72
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(2-Fluorobenzyl)piperidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only