missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 96.0+%, TCI America™
$87.60 - $523.79
Chemical Identifiers
| CAS | 36775-23-2 |
|---|---|
| Molecular Formula | C11H20ClN2O2 |
| MDL Number | MFCD00041858 |
| Synonym | 4-(2-Chloroacetamido)-TEMPO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C1432100MG
|
TCI America
C1432100MG |
100 mg |
Each for $87.60
|
|
|||||
|
C14321G
|
TCI America
C14321G |
1 g |
Each for $523.79
|
|
|||||
Chemical Identifiers
| 36775-23-2 | |
| C11H20ClN2O2 | |
| MFCD00041858 | |
| 4-(2-Chloroacetamido)-TEMPO |
Specifications
| 36775-23-2 | |
| C11H20ClN2O2 | |
| 100 mg | |
| 247.74 | |
| Crystalline Powder |
| 117°C | |
| MFCD00041858 | |
| 4-(2-Chloroacetamido)-TEMPO | |
| ≥96.0% (HPLC) | |
| 4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical |
Safety and Handling
TSCA : Yes