missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Carboxyethyl)benzeneboronic acid, 97%

Catalog No. p-7047062
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1748503 1 g
AAL1748506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1748503 Supplier Thermo Scientific Chemicals Supplier No. L1748503
Only null left

CAS: 166316-48-9 | C9H11BO4 | 193.993 g/mol

4-(2-Carboxyethyl)benzeneboronic acid is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Carboxyethyl)benzeneboronic acid is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 166316-48-9
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.993
MDL Number MFCD01318119
InChI Key VPSARXNVXCRDIV-UHFFFAOYSA-N
Synonym 3-4-boronophenyl propanoic acid, 4-2-carboxyethyl phenylboronic acid, 4-2-carboxyethyl benzeneboronic acid, 3-4-boronophenyl propionic acid, 3-4-dihydroxyboranyl phenyl propanoic acid, acmc-209dty, benzenepropanoic acid,4-borono, 3-4-boronophenyl-propanoic acid, 4-borono-benzenepropanoic acid, 4-2-carboxyethyl phenyl boronic acid
PubChem CID 3863484
IUPAC Name 3-(4-boronophenyl)propanoic acid
SMILES B(C1=CC=C(C=C1)CCC(=O)O)(O)O

Specifications

Melting Point 146°C to 148°C
Quantity 1 g
Beilstein 9198524
Solubility Information Slightly soluble in water.
Formula Weight 193.99
Percent Purity 97%
Chemical Name or Material 4-(2-Carboxyethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only