missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Bromophenyl)pyridine 98.0+%, TCI America™

Catalog No. B5153200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5153200MG 200 mg
B51531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5153200MG Supplier TCI America Supplier No. B5153200MG
Only null left

Chemical Identifiers

CAS 101681-34-9
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.10
MDL Number MFCD18415515
InChI Key GZZVURXUOZUXDO-UHFFFAOYSA-N
PubChem CID 20258453
IUPAC Name 4-(2-bromophenyl)pyridine
SMILES BrC1=CC=CC=C1C1=CC=NC=C1

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 89°C
Quantity 200 mg
Formula Weight 234.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Bromophenyl)pyridine