missing translation for 'onlineSavingsMsg'
Learn More

4-(2-bromoethyl)tetrahydropyran, 97%, Thermo Scientific™

Catalog No. p-3586735
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC67408DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC67408DA Supplier Thermo Scientific Chemicals Supplier No. CC67408DA

Chemical Identifiers

CAS 4677-20-7
Molecular Formula C7H13BrO
Molecular Weight (g/mol) 193.08
MDL Number MFCD09800414
InChI Key WPDAWFCZGSQOPZ-UHFFFAOYSA-N
Synonym 4-2-bromoethyl tetrahydropyran, 4-2-bromoethyl tetrahydro-2h-pyran, 4-2-bromoethyl oxane, 4-2-bromoethyl-tetrahydropyran, 4-2-bromo-ethyl-tetrahydro-pyran, 2h-pyran, 4-2-bromoethyl tetrahydro, 4-2-bromoethyl-tetrahydro-2h-pyran, pubchem16818, ablock ab-11-0011, 2h-pyran,4-2-bromoethyl tetrahydro
PubChem CID 22637012
IUPAC Name 4-(2-bromoethyl)oxane
SMILES BrCCC1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.08
Percent Purity 97%
Chemical Name or Material 4-(2-Bromoethyl)tetrahydropyran
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.