missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Aminoethyl)thiomorpholine 1,1-Dioxide 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 92°C |
| Color | White-Yellow |
| Boiling Point | 182°C |
| Quantity | 5 g |
| Formula Weight | 178.25 |
| Percent Purity | ≥97.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-(2-Aminoethyl)thiomorpholine 1,1-Dioxide |
Safety and Handling
| TSCA | No |