missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)thiomorpholine 1,1-Dioxide 97.0+%, TCI America™

Catalog No. A18031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18031G 1 g
A18035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18031G Supplier TCI America Supplier No. A18031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89937-52-0
Molecular Formula C6H14N2O2S
Molecular Weight (g/mol) 178.25
InChI Key NICIHZYGEQHDPN-UHFFFAOYSA-N
Synonym 2-(1,1-Dioxothiomorpholino)ethylamine
PubChem CID 2760618
IUPAC Name 2-(1,1-dioxo-1,4-thiazinan-4-yl)ethanamine
SMILES C1CS(=O)(=O)CCN1CCN

Specifications

Melting Point 92°C
Color White-Yellow
Boiling Point 182°C
Quantity 1 g
Formula Weight 178.25
Percent Purity ≥97.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Aminoethyl)thiomorpholine 1,1-Dioxide