missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Aminoethyl)pyridine 97.0+%, TCI America™
$262.83 - $872.38
Chemical Identifiers
| CAS | 13258-63-4 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.17 |
| MDL Number | MFCD00038045 |
| InChI Key | IDLHTECVNDEOIY-UHFFFAOYSA-N |
| Synonym | 4-2-aminoethyl pyridine, 4-pyridineethanamine, 2-4-pyridyl ethylamine, 2-pyridin-4-yl-ethylamine, 2-pyridin-4-yl ethan-1-amine, 2-pyridin-4-yl ethanamine, 2-4-pyridinyl ethylamine, 2-pyridin-4-ylethyl amine, 4-pyridine ethaneamine, 2-4-pyridinyl ethanamine |
| PubChem CID | 83275 |
| IUPAC Name | 2-(pyridin-4-yl)ethan-1-amine |
| SMILES | NCCC1=CC=NC=C1 |
Chemical Identifiers
| 13258-63-4 | |
| 122.17 | |
| IDLHTECVNDEOIY-UHFFFAOYSA-N | |
| 83275 | |
| NCCC1=CC=NC=C1 |
| C7H10N2 | |
| MFCD00038045 | |
| 4-2-aminoethyl pyridine, 4-pyridineethanamine, 2-4-pyridyl ethylamine, 2-pyridin-4-yl-ethylamine, 2-pyridin-4-yl ethan-1-amine, 2-pyridin-4-yl ethanamine, 2-4-pyridinyl ethylamine, 2-pyridin-4-ylethyl amine, 4-pyridine ethaneamine, 2-4-pyridinyl ethanamine | |
| 2-(pyridin-4-yl)ethan-1-amine |
Specifications
| 13258-63-4 | |
| C7H10N2 | |
| 5 g | |
| 4-2-aminoethyl pyridine, 4-pyridineethanamine, 2-4-pyridyl ethylamine, 2-pyridin-4-yl-ethylamine, 2-pyridin-4-yl ethan-1-amine, 2-pyridin-4-yl ethanamine, 2-4-pyridinyl ethylamine, 2-pyridin-4-ylethyl amine, 4-pyridine ethaneamine, 2-4-pyridinyl ethanamine | |
| NCCC1=CC=NC=C1 | |
| 122.17 | |
| 122.17 | |
| Liquid |
| 120°C | |
| MFCD00038045 | |
| 2735 | |
| IDLHTECVNDEOIY-UHFFFAOYSA-N | |
| 2-(pyridin-4-yl)ethan-1-amine | |
| 83275 | |
| ≥97.0% (T) | |
| 4-(2-Aminoethyl)pyridine |
Safety and Handling
TSCA : Yes