missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)pyridine, 98%

Catalog No. AC202030010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC202030010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC202030010 Supplier Thermo Scientific Chemicals Supplier No. 202030010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13258-63-4 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13258-63-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00038045
InChI Key IDLHTECVNDEOIY-UHFFFAOYSA-N
Synonym 4-2-aminoethyl pyridine, 4-pyridineethanamine, 2-4-pyridyl ethylamine, 2-pyridin-4-yl-ethylamine, 2-pyridin-4-yl ethan-1-amine, 2-pyridin-4-yl ethanamine, 2-4-pyridinyl ethylamine, 2-pyridin-4-ylethyl amine, 4-pyridine ethaneamine, 2-4-pyridinyl ethanamine
PubChem CID 83275
IUPAC Name 2-pyridin-4-ylethanamine
SMILES NCCC1=CC=NC=C1

Specifications

Density 1.0210g/mL
Boiling Point 112.0°C to 113.0°C (12.0 mmHg)
Flash Point 100°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5403
Linear Formula (C5H4N)CH2CH2NH2
Quantity 1 g
Specific Gravity 1.021
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 4-(2-Aminoethyl)pyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only