missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)morpholine, 98+%

Catalog No. AAB2136530
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2136530 250 g
AAB2136518 50 g
AAB213650B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2136530 Supplier Thermo Scientific Chemicals Supplier No. B2136530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2038-03-1 | C6H14N2O | 130.19 g/mol

4-(2-Aminoethyl)morpholine is used as a ligand and reacts with nickel(II) nitrite to form trans-bis[4-(2-aminoethyl)morpholine]dinitronickel(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Aminoethyl)morpholine is used as a ligand and reacts with nickel(II) nitrite to form trans-bis[4-(2-aminoethyl)morpholine]dinitronickel(II).

Solubility
Miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.

Chemical Identifiers

CAS 2038-03-1
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD00006182
InChI Key RWIVICVCHVMHMU-UHFFFAOYSA-N
Synonym 4-2-aminoethyl morpholine, 2-morpholinoethanamine, 4-morpholineethanamine, 2-morpholinoethylamine, n-aminoethylmorpholine, 2-morpholin-4-yl ethan-1-amine, morpholine, 4-2-aminoethyl, n-2-aminoethyl morpholine, n-2-aminoethylmorpholine, 2-morpholin-4-ylethylamine
PubChem CID 408285
IUPAC Name 2-morpholin-4-ylethanamine
SMILES C1COCCN1CCN

Specifications

Melting Point 23°C to 25°C
Density 0.999
pH 12
Boiling Point 200°C to 203°C
Flash Point 86°C (186°F)
Odor Amine-like
Refractive Index 1.476
Quantity 250 g
UN Number UN2923
Beilstein 104378
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 130.19
Percent Purity ≥98%
Chemical Name or Material 4-(2-Aminoethyl)morpholine
Show More Show Less
Hazard Category H302-H311-H314-H335
Hazard Statement GHS H Statement
H311-H314-H318-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 218-011-6
RTECSNumber QD7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only