missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride 99.0+%, TCI America™

Catalog No. A2215100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2215100MG 100 mg
A22151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2215100MG Supplier TCI America Supplier No. A2215100MG

Chemical Identifiers

CAS 30827-99-7
Molecular Formula C8H11ClFNO2S
Molecular Weight (g/mol) 239.689
MDL Number MFCD00132962
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
PubChem CID 186136
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl

Specifications

Melting Point 183°C
Color White-Yellow
Quantity 100 mg
UN Number 3261
Formula Weight 239.69
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride [for Biochemical Research]
TSCA No
Recommended Storage Freezer