Learn More
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, 97%
CAS: 30827-99-7 | C8H11ClFNO2S | 239.689 g/mol
Supplier: Thermo Scientific Chemicals H2647303
Description
AEBSF is extensively used in studies aiming to describe cholesterol regulatory genes due to its potent ability to inhibit Site-1-protease (S1P). This serine protease, located in the Golgi apparatus, is responsible for activating the sterol regulatory element-binding proteins. By selectively inhibiting S1P, AEBSF can be used to characterize the downstream result of SREBP inhibition and its influence on cholesterol regulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAEBSF is extensively used in studies aiming to describe cholesterol regulatory genes due to its potent ability to inhibit Site-1-protease (S1P). This serine protease, located in the Golgi apparatus, is responsible for activating the sterol regulatory element-binding proteins. By selectively inhibiting S1P, AEBSF can be used to characterize the downstream result of SREBP inhibition and its influence on cholesterol regulation.
Solubility
Soluble in water 50 mg/mL or 100% ethanol.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
Specifications
| 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride | |
| ∼184°C (decomposition) | |
| MFCD00132962 | |
| UN3261 | |
| Moisture sensitive | |
| Soluble in water 50mg/mL or 100% ethanol. | |
| C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl | |
| 239.689 | |
| 239.69 |
| 30827-99-7 | |
| C8H11ClFNO2S | |
| 1 g | |
| 7604627 | |
| 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride | |
| WRDABNWSWOHGMS-UHFFFAOYSA-N | |
| 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride | |
| 186136 | |
| 97% |
Chemical Identifiers
| 30827-99-7 | |
| 239.689 | |
| WRDABNWSWOHGMS-UHFFFAOYSA-N | |
| 186136 | |
| C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl |
| C8H11ClFNO2S | |
| MFCD00132962 | |
| 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride | |
| 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H500
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only