missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)-1-benzylpiperidine 98.0+%, TCI America™

Catalog No. p-7117490
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15175G 5 g
A151725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15175G Supplier TCI America Supplier No. A15175G
Only null left

Chemical Identifiers

CAS 86945-25-7
Molecular Formula C14H22N2
Molecular Weight (g/mol) 218.344
MDL Number MFCD02093423
InChI Key OUYRPOHWEJUTCQ-UHFFFAOYSA-N
Synonym 2-(1-Benzyl-4-piperidyl)ethylamine
PubChem CID 1867002
IUPAC Name 2-(1-benzylpiperidin-4-yl)ethanamine
SMILES C1CN(CCC1CCN)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 5 g
Formula Weight 218.34
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(2-Aminoethyl)-1-benzylpiperidine