missing translation for 'onlineSavingsMsg'
Learn More

4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile, 98%

Catalog No. AAH6117806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6117806 5 g
AAH6117803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6117806 Supplier Thermo Scientific Chemicals Supplier No. H6117806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 119516-86-8 | C13H12N2 | 196.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119516-86-8
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.253
InChI Key FNDFKQYZEDOHRC-UHFFFAOYSA-N
Synonym 1-4-cyanophenyl-2,5-dimethylpyrrole, 4-2,5-dimethylpyrrol-1-yl benzonitrile, 4-2,5-dimethyl-1h-pyrrol-1-yl benzonitrile, 2,5-dimethyl-1-4-cyanophenyl pyrrole, n-4'-cyanophenyl-2,5-dimethylpyrrole, 4-2,5-dimethyl-1-pyrrolyl benzonitrile, benzonitrile,4-2,5-dimethyl-1h-pyrrol-1-yl
PubChem CID 3787857
IUPAC Name 4-(2,5-dimethylpyrrol-1-yl)benzonitrile
SMILES CC1=CC=C(N1C2=CC=C(C=C2)C#N)C

Specifications

Quantity 5 g
UN Number UN3439
Formula Weight 196.25
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only